missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen sulfate, fused, ACS, mixture of potassium pyrosulfate, K{2}S{2}O{7}, and potassium bisulfate, KHSO{4}, Thermo Scientific Chemicals

Catalog Number p-4824892
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
MFCD00066385
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
MFCD00066385

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It is also used as a disintegrating agent in analytical chemistry. Further, it is used to make fertilizers and in the preparation of potassium persulfate. In addition to this, it is used as a catalyst in organic preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 214°C
Color White
Physical Form Fused Powder
Quantity 100 g
Solubility Information Soluble in water,isopropyl alcohol and acetone.
Formula Weight 136.17
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White fused powder
Chemical Name or Material Potassium hydrogen sulfate, fused
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'