missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical™
200.44 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Description
Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 877-24-7, White, Melting Point: 295 deg.C, Molecular Formula: C8H5KO4, Molecular Weight: 204.222, Packaging: Plastic powder jar, pH: 3.8 to 4.0
Specifications
Melting Point | 295°C |
Color | White |
pH | 3.8 to 4.0 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 204.22g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'