missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium hydrogen carbonate, 99.5%, for biochemistry

Catalog Number p-4813612
28.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
298-14-6
CHKO3
100.11
MFCD00011402
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
516893
CHEBI:81862
[K+].OC([O-])=O
This item is not returnable. View return policy
298-14-6
CHKO3
100.11
MFCD00011402
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
516893
CHEBI:81862
[K+].OC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 292.0°C
Quantity 500 g
Linear Formula KHCO3
Merck Index 15773
Solubility Information Clear colorless
IUPAC Name potassium hydrogen carbonate
Formula Weight 100.12
Percent Purity 99.5%
Grade Biochemical
Packaging Plastic Bottle
Chemical Name or Material Potassium hydrogen carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'