missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydride, 25 to 35 wt.%, dispersion in mineral oil, ACROS Organics™

Brand:  Acros Organics 387960250

Code : 29

Additional Details : CAS Number : 7693-26-7 Weight : 0.07500kg Transport : UN number : 3129 Chem class : 1b Pack group : I

Quantity 25g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10535443

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Potassium hydride
HK
40.11
8042-47-5
KH

Specifications

Potassium hydride
Dispersion of Powder In Mineral Oil
KH
25.0 to 35.0 wt%
Plastic bottle
316.0°C
25g
8042-47-5
HK
40.11
1.5400g/mL
1.54
Grayish beige
Videos
SDS