missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydride, 25 to 35 wt.%, dispersion in mineral oil, ACROS Organics™

Brand:  Acros Organics 387961000

Code : 29

Additional Details : CAS Number : 7693-26-7 Weight : 0.15000kg Transport : UN number : 3129 Chem class : 1b Pack group : I

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10026122

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Potassium hydride
HK
40.11
8042-47-5
KH

Specifications

Potassium hydride
Dispersion of Powder In Mineral Oil
KH
25.0 to 35.0 wt%
Plastic bottle
316.0°C
100g
8042-47-5
HK
40.11
1.5400g/mL
1.54
Grayish beige
Videos
SDS