missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium hexanitrorhodate(III), Premion™, 99.99% (metals basis), Rh 20.2% min

Brand:  Alfa Aesar™ 040410.03

Code : NEW

Additional Details : CAS Number : 17712-66-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11384008

  • £100.81 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Potassium hexanitrorhodate(III)
Premion™
496.23
17712-66-2
K3Rh(NO2)6
MFCD00049776

Specifications

Potassium hexanitrorhodate(III)
Premion™
99.99%
K3Rh(NO2)6
MFCD00049776
White to yellow
17712-66-2
Powder
(metals basis)
496.23
Rh 20.2% min.
1g
Videos
SDS