Learn More
Thermo Scientific Alfa Aesar Potassium hexanitrorhodate(III), Premion™, 99.99% (metals basis), Rh 20.2% min
Brand: Thermo Scientific Alfa Aesar 040410.06
Potassium hexanitrorhodate(III), Premion™, 99.99% (metals basis), Rh 20.2% min
Code : NEW
Additional Details : CAS Number : 17712-66-2 Weight : 0.00500kg