missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexahydroxyplatinate(IV), Premion™, 99.95% (metals basis), Pt 51.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Potassium hexahydroxyplatinate(IV) is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 375.34 |
Percent Purity | 99.95% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Pt 51.5% min. |
Assay | (metals basis) |
Chemical Name or Material | Potassium hexahydroxyplatinate(IV) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'