missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexafluorozirconate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Employed in the manufacture of metallic zirconium. Used in the production of aluminum magnesium alloy, ceramics and glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 500 g |
UN Number | UN3288 |
Merck Index | 14,7639 |
Solubility Information | Slightly soluble in cold water, soluble in hot water |
Formula Weight | 283.41 |
Percent Purity | 99% |
Odor | Odorless |
Density | 3.48 g/mL |
Chemical Name or Material | Potassium hexafluorozirconate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'