missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexafluorophosphate, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number 10713391
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
17084-13-8
F6KP
184.07
MFCD00011412
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
23688904
potassium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[K+]
17084-13-8
F6KP
184.07
MFCD00011412
YZDGRYDIGCWVND-UHFFFAOYSA-N
potassium hexafluorophosphate, potassium fluophosphate, potassium hexafluorophosphate v, unii-88w436pdi9, monopotassium hexafluorophosphate, potassium hexfluorophosphate, potassium hexafluorophosphate min, phosphate 1-, hexafluoro-, potassium, potassium fluorophosphate, kpf6
23688904
potassium;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[K+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 575.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 98.5% min
Linear Formula KPF6
Solubility Information Solubility in water: 93g/L (25°C)
Formula Weight 184.07
Percent Purity 99%
Grade Extra Pure
Loss on Drying 0.1% max. (100°C)
Packaging Plastic bottle
Chemical Name or Material Potassium hexafluorophosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'