missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexafluorophosphate, 95% min, Thermo Scientific Chemicals
42.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Potassium hexafluorophosphate is a soluble neutral salt used for synthesis. It is a source of the hexafluorophosphate anion, a non-coordinating anion that confers lipophilicity to its salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 575°C |
Color | White |
pH | 3.0 to 7.0 |
Physical Form | Crystalline |
Quantity | 250 g |
UN Number | UN3260 |
Solubility Information | Soluble in water (83.5mg/ml at 20°C). |
Formula Weight | 184.07 |
Percent Purity | ≤95% |
Odor | Odorless |
Show More |
Safety and Handling
Product Identifier
- Potassium hexafluorophosphate
- Danger
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 4
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'