missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium hexafluoroantimonate, 98% min

Brand:  Alfa Aesar™ 040146.09

Code : NEW

Additional Details : CAS Number : 16893-92-8 Weight : 0.01000kg Transport : UN number : 1549 Chem class : 1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 11329297

  • £51.62 / 10g


Estimated Shipment date
from Supplier 13-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium hexafluoroantimonate
274.84
UN1549
16893-92-8
MFCD00043080

Specifications

Potassium hexafluoroantimonate
Powder
274.84
UN1549
1505°C
10g
16893-92-8
≤98%
MFCD00043080
Odorless
846°C
Videos
SDS