missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hexacyanoferrate(III), Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
Specifications
Color | Orange to Red |
pH | 6 |
Physical Form | Crystalline Solid |
Quantity | 250 g |
Formula Weight | 329.26g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'