missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(III), Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/4840/53

Code : 21

Additional Details : CAS Number : 13746-66-2 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 10172150

  • £129.00 / 500g

In stock, ready to ship: 4

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

13746-66-2
329.248
BYGOPQKDHGXNCD-UHFFFAOYSA-N
26250
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[K+].[K+].[K+].[Fe+3]
C6FeK3N6
11392
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferriccyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
tripotassium;iron(3+);hexacyanide

Specifications

13746-66-2
11392
BYGOPQKDHGXNCD-UHFFFAOYSA-N
tripotassium;iron(3+);hexacyanide
26250
Crystalline Solid
Orange to Red
500g
C6FeK3N6
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferriccyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[K+].[K+].[K+].[Fe+3]
329.248
329.26g/mol
Plastic powder jar
6
Videos
SDS Documents
Certificates