missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexacyanocobaltate(III), 95%, Thermo Scientific Chemicals
67.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Yellow |
Physical Form | Crystalline Solid |
Quantity | 100 g |
Assay Percent Range | 95% |
Linear Formula | K3[Co(CN)6] |
Merck Index | 15, 7758 |
Solubility Information | Solubility in water: freely soluble. Other solubilities: insoluble in alcohol |
Formula Weight | 332.34 |
Percent Purity | 95% |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Potassium hexacyanocobaltate(III)
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'