missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexacyanocobaltate(III), 90+%, Thermo Scientific Chemicals
53.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
It is applied in formation of a xerogel via a cyanogel, the gel has potential use in solid-state gas sensors. It is also used as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Brown to Yellow |
Physical Form | Crystalline |
Quantity | 50 g |
Linear Formula | K3[Co(CN)6] |
Merck Index | 14,7637 |
Solubility Information | Soluble in water and acetic acid solutions; Insoluble in alcohol |
Formula Weight | 332.33 |
Percent Purity | ≥90% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'