missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 23% min, Thermo Scientific Chemicals

Catalog Number p-7059413
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
25443-63-4
K3RuCl6
MFCD00011390
ruthenium 6+ hexachloride, tripotassium hexachlororuthenate, ruthenium hexachloride, ruthenium chloride rucl6 , oc-6-11, ruthenate 3-, tripotassium, oc-6-11
This item is not returnable. View return policy
25443-63-4
K3RuCl6
MFCD00011390
ruthenium 6+ hexachloride, tripotassium hexachlororuthenate, ruthenium hexachloride, ruthenium chloride rucl6 , oc-6-11, ruthenate 3-, tripotassium, oc-6-11

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 1 g
Formula Weight 431.09
Percent Purity 99.99%
Grade Premion™
Concentration or Composition (by Analyte or Components) Ru 23% min
Assay (metals basis)
Chemical Name or Material Potassium hexachlororuthenate (III)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'