missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium hexachloropalladate(IV), Pd 26.3% min

Brand:  Alfa Aesar™ 011884.14

Code : NEW

Additional Details : CAS Number : 16919-73-6 Weight : 0.02500kg Transport : UN number : 3077 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11371968

  • £721.10 / 25g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium hexachloropalladate(IV)
397.32
UN3077
16919-73-6
MFCD00043081

Specifications

Potassium hexachloropalladate(IV)
Crystalline
MFCD00043081
Pd 26.3% min.
Odorless
Red
16919-73-6
397.32
UN3077
2.738g/mL
Hygroscopic
25g
Videos
SDS