missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexachloroosmate(IV), Os 38.7% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
5 g
Description
Potassium hexachloroosmate(IV) used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Red |
Physical Form | Powder |
Quantity | 1 g |
Linear Formula | K2OsCl6 |
Merck Index | 14,7635 |
Solubility Information | Soluble in water; Sparingly soluble in alcohol |
Formula Weight | 481.12 |
Concentration or Composition (by Analyte or Components) | Os 38.7% min. |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'