missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexabromotellurate(IV), 99.99% (metals basis), Thermo Scientific™

Catalog Number p-7059152
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
16986-18-8
Br6K2Te
685.221
MFCD03427026
HMFRPYOIJYYQFO-UHFFFAOYSA-L
potassium hexabromotellurate iv
121233754
[K+].[K+].[Br-].[Br-].Br[Te](Br)(Br)Br
This item is not returnable. View return policy
16986-18-8
Br6K2Te
685.221
MFCD03427026
HMFRPYOIJYYQFO-UHFFFAOYSA-L
potassium hexabromotellurate iv
121233754
[K+].[K+].[Br-].[Br-].Br[Te](Br)(Br)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 5 g
UN Number UN3284
Formula Weight 685.22
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Potassium hexabromotellurate (IV)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'