missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium hexabromoosmate, Premion™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 039507.04

Code : NEW

Additional Details : CAS Number : 16903-69-8 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11398308

  • £375.03 / 2g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium hexabromoosmate
Premion™
747.86
16903-69-8
K2OsBr6
MFCD00049658

Specifications

Potassium hexabromoosmate
Premion™
99.99%
K2OsBr6
MFCD00049658
2g
16903-69-8
Powder
(metals basis)
747.86
Red to black
Videos
SDS