missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium heptafluorotantalate(V), 99.7%

Brand:  Alfa Aesar™ 040316.30

Code : NEW

Additional Details : CAS Number : 16924-00-8 Weight : 0.25000kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 250g

 View more versions of this product

Product Code. 11313878

  • £795.00 / 250g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium heptafluorotantalate(V)
392.14
UN3288
16924-00-8
MFCD00042369
Tantalum potassium fluoride

Specifications

Potassium heptafluorotantalate(V)
Powder
392.14
UN3288
Decomposes in water; Slightly soluble in HF
250g
16924-00-8
99.7%
MFCD00042369
Tantalum potassium fluoride
4.560g/mL
Videos
SDS