missing translation for 'onlineSavingsMsg'
Learn More

Potassium heptafluoroniobate(V), 99.5%, Thermo Scientific Chemicals

Catalog Number p-7080288
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
16924-03-1
K2NbF7
MFCD00042368
This item is not returnable. View return policy
16924-03-1
K2NbF7
MFCD00042368

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 10 g
UN Number UN3288
Formula Weight 304.1
Percent Purity 99.5%
Sensitivity Hygroscopic
Chemical Name or Material Potassium heptafluoroniobate (V)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'