missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride dihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific™

Catalog Number p-7048549
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
13455-21-5
FH4KO2
94.13
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
23708904
potassium dihydrate fluoride
O.O.[F-].[K+]
This item is not returnable. View return policy
13455-21-5
FH4KO2
94.13
MFCD00149929
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
23708904
potassium dihydrate fluoride
O.O.[F-].[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 41°C
Physical Form Powder
Quantity 10 g
UN Number UN1812
Merck Index 14,7632
Formula Weight 94.13 (58.10 Anhydrous)
Percent Purity 99.995%
Grade Puratronic™
Odor Odorless
Refractive Index 1.352
Sensitivity Hygroscopic
Density 2.5g/mL
Assay (metals basis)
Chemical Name or Material Potassium fluoride dihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'