missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390831
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7789-23-3
FK
58.10
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
522689
CHEBI:66872
[F-].[K+]
This item is not returnable. View return policy
7789-23-3
FK
58.10
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
522689
CHEBI:66872
[F-].[K+]

High quality chemicals for laboratory and specialized industrial use

Quantity 500 g
IUPAC Name potassium;fluoride
Formula Weight 58.1
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.48kg
Chemical Name or Material Potassium Fluoride, Anhydrous
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'