missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Boiling Point | 1505.0°C |
Melting Point | 860.0°C |
Color | White |
Physical Form | Crystalline Powder, Crystals and/or Chunks |
Quantity | 2.5 kg |
Assay Percent Range | 99% |
Linear Formula | KF |
Fieser | 01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282 |
Merck Index | 15, 7753 |
Solubility Information | Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid, insoluble in alcohol unless water is present |
Show More |
Safety and Handling
Product Identifier
- Potassium fluoride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'