missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferricyanide, ≥99.0%, (ACS Reagent Grade), MP Biomedicals
Click to view available options
Quantity:
100 g
500 g
Specifications
Quantity | 100 g |
Assay Percent Range | ≥99.0% |
Formula Weight | 329.5 |
Grade | ACS Reagent |
Chemical Name or Material | Potassium Ferricyanide |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : potassium ferricyanide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'