missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium ferricyanide, 98%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Color | Orange to Red |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 1 kg |
Assay Percent Range | 97.5% min. (Iodometry) |
Linear Formula | K3Fe(CN)6 |
Fieser | 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255 |
Merck Index | 15, 7751 |
Solubility Information | Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol |
IUPAC Name | tripotassium hexacyanoirontris(ylium) |
Formula Weight | 329.26 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'