missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium dinitrosulfatopalladate(II), solution, Pd 5% (cont. Pd)

Brand:  Alfa Aesar™ 012629.NB

Code : NEW

Additional Details : CAS Number : 67859-45-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11460300

  • £286.20 / 5g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Potassium dinitrosulfatopalladate(II)
Solution
372.69
67859-45-4
K2Pd(NO2)2SO4
MFCD00064669

Specifications

Potassium dinitrosulfatopalladate(II)
67859-45-4
K2Pd(NO2)2SO4
MFCD00064669
5g
Solution
Liquid
372.69
Pd 5% (cont. Pd)
Videos
SDS