missing translation for 'onlineSavingsMsg'
Learn More

Potassium dihydrogen phosphate, Puratronic™, 99.999% (metals basis), Si 10ppm max, Thermo Scientific Chemicals

Catalog Number p-7048548
Click to view available options
Quantity:
25 g
100 g
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
516951
CHEBI:63036
[K+].OP(O)([O-])=O
7778-77-0
H2KO4P
136.08
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
516951
CHEBI:63036
[K+].OP(O)([O-])=O

Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 253°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Linear Formula KH2PO4
Merck Index 14,7659
Solubility Information Soluble in water; slightly soluble in ethanol .
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Percent Purity 99.999%
Grade Puratronic™
Concentration or Composition (by Analyte or Components) Si 10ppm max.
Sensitivity Hygroscopic
Density 2.338 g/mL
Assay (metals basis)
Chemical Name or Material Potassium dihydrogen phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'