missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium dichromate, 99.5%, extra pure, Thermo Scientific Chemicals
139.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Boiling Point | 500.0°C |
Melting Point | 398.0°C |
Color | Orange to Orange-Red |
Physical Form | Crystals |
Quantity | 5 kg |
Assay Percent Range | 99.4% min. (Iodometry) |
Linear Formula | K2Cr2O7 |
Fieser | 08,410; 09,383; 10,324; 12,405 |
Merck Index | 15, 7748 |
Solubility Information | Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'