missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate, 0.025N Standardized Solution, Thermo Scientific Chemicals

Catalog Number p-7050101
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7778-50-9
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7778-50-9
Cr2K2O7
294.182
MFCD00011367
KMUONIBRACKNSN-UHFFFAOYSA-N
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
24502
CHEBI:53444
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Linear Formula K2Cr2O7
Concentration 0.025N
Formula Weight 294.18
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Potassium dichromate, Standardized Solution

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'