missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, Technical grade, Honeywell Fluka™

Brand:  Honeywell Fluka 11813-1KG

Code : NEW

Additional Details : CAS Number : 151-50-8 Weight : 1.00000kg Transport : UN number : 1680 Chem class : 1 Pack group : I

Quantity 1kg

 View more versions of this product

Product Code. 15641080

  • £40.65 / 1kg


Estimated Shipment date
from Supplier 26-01-2018
Add to basket

Specifications

Chemical Identifiers

3593645
Potassium Cyanide
65.12g/mol
KCN
1680
151-50-8
≥96%
Technical grade
MFCD00011397

Specifications

Potassium Cyanide
Technical grade
KCN
MFCD00011397
3593645
151-50-8
≥96%
65.12g/mol
1680
1kg
Videos
SDS