missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, ACS Reagent, ≥96.0%, Honeywell Fluka™

Brand:  Honeywell Fluka 207810-25G

Code : NEW

Additional Details : CAS Number : 151-50-8 Weight : 0.02500kg Transport : UN number : 1680 Chem class : 6.1 Pack group : I

Packaging Poly Bottle
Quantity 25g

 View more versions of this product

Product Code. 15641790

  • £14.42 / 25g


Estimated Shipment date
from Supplier 29-08-2018
Add to basket

Specifications

Chemical Identifiers

151-50-8
65.116
NNFCIKHAZHQZJG-UHFFFAOYSA-N
CKN
MFCD00011397
9032

Specifications

Potassium Cyanide
ACS Reagent
CKN
Granular Powder
UN1680
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium;cyanide
9032
ca. 750kg/cu.m
Poly Bottle
1.8 hPa at 634.5°C
White
25g
151-50-8
≥96.0%
KCN
MFCD00011397
3593645
[C-]#N.[K+]
65.116
65.12g/mol
1.56g/cu.cm
Soluble in water
Solid
634°C (lit.)
Videos
SDS