missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, 97%, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-43050
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
151-50-8
CKN
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
151-50-8
CKN
MFCD00011397
NNFCIKHAZHQZJG-UHFFFAOYSA-N
potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
9032

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 97%
Linear Formula CKN
Fieser 05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22
Merck Index 15, 7747
SMILES [C-]#N.[K+]
IUPAC Name potassium;cyanide
Molecular Weight (g/mol) 65.116
Formula Weight 65.12
Percent Purity ≥96.0%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Potassium cyanide, 96+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'