missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate, Puriss., ≥99%, Honeywell Fluka™

Brand:  Honeywell Fluka 12249-1KG

Code : NEW

Additional Details : CAS Number : 7789-00-6 Weight : 1.00000kg Transport : UN number : 3087 Chem class : 5.1 Pack group : II

Quantity 1kg

 View more versions of this product

Product Code. 15641110

  • £88.02 / 1kg


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Specifications

Chemical Identifiers

7789-00-6
≥99%
K2CrO4
3288
Potassium Chromate
puriss.
MFCD00011368
6.7 (vs air)

Specifications

Potassium Chromate
7789-00-6
≥99%
K2CrO4
3288
[O-][Cr](=O)(=O)[O-].[K+].[K+]
194.189
CHEBI:75249
1kg
194.19g/mol
puriss.
CrK2O4
MFCD00011368
XMXNVYPJWBTAHN-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)chromium
24597
6.7 (vs air)
Videos