missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chromate, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10181910
66.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


66.30 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7789-00-6
CrK2O4
194.19
MFCD00011368
XMXNVYPJWBTAHN-UHFFFAOYSA-N
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
24597
CHEBI:75249
dipotassium;dioxido(dioxo)chromium
[K+].[K+].[O-][Cr]([O-])(=O)=O
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
7789-00-6
CrK2O4
194.19
MFCD00011368
XMXNVYPJWBTAHN-UHFFFAOYSA-N
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
24597
CHEBI:75249
dipotassium;dioxido(dioxo)chromium
[K+].[K+].[O-][Cr]([O-])(=O)=O

CrK2O4, CAS Number-7789-00-6, neutral potassium chromate, dipotassium monochromate, chromate of potass, caswell no. 687, bipotassium chromate, potassium chromate, dipotassium chromate, chromate of potash, potassium chromate vi, tarapacaite, 250g, CHEBI:75249, Yellow, 194.2g/mol

TRUSTED_SUSTAINABILITY
Melting Point 975°C
Color Yellow
pH 8.6 to 9.8
Physical Form Solid
Quantity 250 g
Formula Weight 194.2g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium chromate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H340-May cause genetic defects.
  • H350i-May cause cancer by inhalation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'