missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
Description
CrK2O4, CAS Number-7789-00-6, neutral potassium chromate, dipotassium monochromate, chromate of potass, caswell no. 687, bipotassium chromate, potassium chromate, dipotassium chromate, chromate of potash, potassium chromate vi, tarapacaite, 250g, CHEBI:75249, Yellow, 194.2g/mol
Specifications
Melting Point | 975°C |
Color | Yellow |
pH | 8.6 to 9.8 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 194.2g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'