missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Extra Pure, SLR, Eur. Ph., Fisher Chemical™
Click to view available options
Quantity:
500 g
Description
ClK, CAS Number-7447-40-7, enseal, potassium chloride kcl, kalitabs, potassium chloride, muriate of potash, chlorvescent, klotrix, klor-con, slow-k, sylvite, 500g, 1420 deg.C, CHEBI:32588, White, 74.54g/mol, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 770 deg.C, 11360, 74.548
Specifications
Boiling Point | 1420°C |
Melting Point | 770°C |
Color | White |
pH | 6 |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | potassium chloride |
Formula Weight | 74.54g/mol |
Grade | Extra Pure |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'