missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium chloride crystal optic rectangle, 41mm x 23mm x 6mm, unpolished

Brand:  Alfa Aesar™ 038405.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15476205

  • £35.30 / Each


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Specifications

Potassium chloride crystal optic rectangle
41mm x 23mm x 6mm, Unpolished
MFCD00011360
1 each
Videos
SDS