missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, (drilled), polished both sides

Brand:  Alfa Aesar™ 038474.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15473655

  • £31.20 / Each


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Specifications

Potassium chloride crystal optic rectangle
38.5mm x 19.5mm x 4mm, (drilled), polished both sides
MFCD00011360
1 each
Videos
SDS