missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium chloride crystal optic disc, 32mm x 3mm, polished both sides

Brand:  Alfa Aesar™ 038421.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15496215

  • £33.70 / Each


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Specifications

Potassium chloride crystal optic disc
32mm x 3mm, Polished both sides
MFCD00011360
1 each
Videos
SDS