missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 3040-05

Code : NEW

Additional Details : CAS Number : 7447-40-7 Weight : 2.50000kg

Quantity 2.5kg
Packaging Poly Bottle

 View more versions of this product

Product Code. 15119145

  • £221.00 / 2.50kg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

Potassium Chloride
Baker Analyzed™ ACS Reagent
74.55
7447-40-7
KCl

Specifications

Potassium Chloride
Baker Analyzed™ ACS Reagent
KCl
Poly Bottle
2.5kg
7447-40-7
Crystals
74.55
1L = 1.99kg
Videos