missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Catalog Number p-7048830
17.71 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.


12.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
[Cl-].[K+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
[Cl-].[K+]

In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy

Potassium chloride is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins. It acts as a fire extinguishing agent in portable and wheeled fire extinguishers. It may be used as a softener unit in household water, which is an alternative to sodium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy
TRUSTED_SUSTAINABILITY
Boiling Point 1500°C (sublimation)
Melting Point 773°C
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99.0 to 100.5%
Merck Index 14,7621
Solubility Information Soluble in water,glycerol and alkalies; Slightly soluble in alcohol and insoluble in ether.
IUPAC Name potassium chloride
Formula Weight 74.55
Percent Purity 99.0 to 100.5%
Grade ACS
Odor Odorless
Refractive Index 1.49
Sensitivity Hygroscopic
Density 1.984 g/mL
Chemical Name or Material Potassium chloride, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'