missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals
139.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
120.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
120.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysis
Specifications
Boiling Point | 1420.0°C |
Melting Point | 770.0°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 5 kg |
Assay Percent Range | 99.0 to 100.5% (on dried substance) (Argentometry) |
Linear Formula | KCl |
Fieser | 06,48 |
Merck Index | 15,7742 |
Solubility Information | Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in ether and acetone |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'