missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, ≥99.0%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Boiling Point | 1413°C (1013hPa) |
Melting Point | 772°C |
Color | Colorless |
pH | 5 to 8 (at 20°C) |
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | KCl |
UN Number | NONH for all modes of transport |
Formula Weight | 74.55g/mol |
Percent Purity | ≥99.0% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'