missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, ≥99.0%, Reagent Grade, Honeywell™

Catalog Number 15678030
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]
This item is not returnable. View return policy
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
potassium;chloride
[Cl-].[K+]

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Boiling Point 1413°C (1013hPa)
Melting Point 772°C
Color Colorless
pH 5 to 8 (at 20°C)
Physical Form Solid
Quantity 5 kg
Linear Formula KCl
UN Number NONH for all modes of transport
Formula Weight 74.55g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'