missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
4 L
Description
Potassium chloride 0.010M solution is used as conductive standard and as electrolyte in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 L |
Color | Colorless |
Assay Percent Range | 0.010M |
IUPAC Name | potassium;chloride |
Formula Weight | 74.55 |
Concentration or Composition (by Analyte or Components) | Potassium chloride: 0.075%; Water: 99.925% |
Odor | Odorless |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Density | 1g/cm3 at 20°C |
Solubility | Miscible with water |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'