missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium chloride, 0.00017M, Conductivity Standard

Brand:  Alfa Aesar™ 043444.K7

Code : NEW

Additional Details : CAS Number : 7447-40-7 Weight : 0.01000kg

Quantity 4L

 View more versions of this product

Product Code. 11305757

  • £75.00 / 4 L


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Potassium chloride Conductivity Standard
25μmho at 25°C
74.55
7447-40-7
KCl
MFCD00011360

Specifications

Potassium chloride Conductivity Standard
7447-40-7
KCl
74.55
Odorless
25μmho at 25°C
Liquid
0.00017M
MFCD00011360
4L
Videos
SDS