missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate sesquihydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals

Catalog Number p-7048593
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
6381-79-9
C2H6K4O9
330.45
MFCD00149925
WWVGGBSGYNQKCY-UHFFFAOYSA-J
tetrapotassium trihydrate dicarbonate
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
This item is not returnable. View return policy
6381-79-9
C2H6K4O9
330.45
MFCD00149925
WWVGGBSGYNQKCY-UHFFFAOYSA-J
tetrapotassium trihydrate dicarbonate
O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

For Manufacturing of Soaps, Pigments, Printing Ink

Potassium carbonate sesquihydrate is used for manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts and as an analytical lab reagent. It is also useful as a reagent for removing carbon dioxide from ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent
TRUSTED_SUSTAINABILITY
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.5-101.0%
Merck Index 14,7670
Solubility Information Soluble in water; Practically insoluble in alcohol
Formula Weight 165.23 (138.21 Anhydrous)
Percent Purity 98.5 to 101.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Potassium carbonate sesquihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'